Anti-glutathione S transferase Omega 1 Rabbit monoclonal antibody unconjugated [clone: EPR8371]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131262-100EA 3764.76 DKK
ABCAAB131262-100
Anti-glutathione S transferase Omega 1 Rabbit monoclonal antibody unconjugated [clone: EPR8371]
Antistoffer
Type: Primary
Antigen: glutathione S transferase Omega 1
Clonality: monoclonal
Clone: EPR8371
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR