Anti-PDCD7 Rabbit Monoclonal Antibody [clone: EPR8231]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131258-100EA 3800.38 DKK
ABCAAB131258-100
Anti-PDCD7 Rabbit Monoclonal Antibody [clone: EPR8231]
Antistoffer
Type: Primary
Antigen: PDCD7 (programmed cell death 7)
Clonality: monoclonal
Clone: EPR8231
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR