Anti-DNPH1 Rabbit Monoclonal Antibody [clone: EPR8686]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131252-100EA 6020 DKK
ABCAAB131252-100
Anti-DNPH1 Rabbit Monoclonal Antibody [clone: EPR8686]
Antistoffer
Type: Primary
Antigen: DNPH1 (2'-deoxynucleoside 5'-phosphate N-hydrolase 1)
Clonality: monoclonal
Clone: EPR8686
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR