Anti-G3BP1 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131243-100EA 4174.79 DKK
ABCAAB131243-100
Anti-G3BP1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: G3BP1 (GTPase activating protein (SH3 domain) binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR