Anti-SNAP23 Rabbit Monoclonal Antibody [clone: EPR8538]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131242-100EA 7210 DKK
ABCAAB131242-100
Anti-SNAP23 Rabbit Monoclonal Antibody [clone: EPR8538]
Antistoffer
Type: Primary
Antigen: SNAP23 (synaptosomal-associated protein, 23kDa)
Clonality: monoclonal
Clone: EPR8538
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR