Anti-ESD Rabbit Monoclonal Antibody [clone: EPR8446]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB131211-100EA 6010 DKK
ABCAAB131211-100
Anti-ESD Rabbit Monoclonal Antibody [clone: EPR8446]
Antistoffer
Type: Primary
Antigen: ESD (esterase D)
Clonality: monoclonal
Clone: EPR8446
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu