Anti-NCLN Rabbit Monoclonal Antibody [clone: EPR8528]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131159-100EA 5880 DKK
ABCAAB131159-100
Anti-NCLN Rabbit Monoclonal Antibody [clone: EPR8528]
Antistoffer
Type: Primary
Antigen: NCLN (nicalin)
Clonality: monoclonal
Clone: EPR8528
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR