Anti-COX7A2 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131143-50EA 6150 DKK
ABCAAB131143-50
Anti-COX7A2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: COX7A2 (cytochrome c oxidase subunit VIIa polypeptide 2 (liver))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR