Anti-GPR101 Rabbit polyclonal antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131117-50EA 4174.79 DKK
ABCAAB131117-50
Anti-GPR101 Rabbit polyclonal antibody
Type: Primary
Antigen: GPR101 (G protein-coupled receptor 101)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR