Anti-ATF2 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131106-100EA 4100.24 DKK
ABCAAB131106-100
Anti-ATF2 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR