Anti-GSK3B Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131097-100EA 4286.61 DKK
ABCAAB131097-100
Anti-GSK3B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR