Anti-SLC22A6 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131087-100EA 6480 DKK
ABCAAB131087-100
Anti-SLC22A6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC22A6 (solute carrier family 22 (organic anion transporter), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR