Anti-ITGA3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131055-100EA 3578.39 DKK
ABCAAB131055-100
Anti-ITGA3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ITGA3 (integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR