Anti-HDGF Rabbit Monoclonal Antibody [clone: EPR7899]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131046-100EA 3913.86 DKK
ABCAAB131046-100
Anti-HDGF Rabbit Monoclonal Antibody [clone: EPR7899]
Antistoffer
Type: Primary
Antigen: HDGF (hepatoma-derived growth factor)
Clonality: monoclonal
Clone: EPR7899
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR