Anti-IL12A Rabbit Monoclonal Antibody [clone: EP5737]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131039-100EA 3839.31 DKK
ABCAAB131039-100
Anti-IL12A Rabbit Monoclonal Antibody [clone: EP5737]
Antistoffer
Type: Primary
Antigen: IL12A (interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35))
Clonality: monoclonal
Clone: EP5737
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR