Anti-JMJD1C Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB130922-100EA 6450 DKK
ABCAAB130922-100
Anti-JMJD1C Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: JMJD1C (jumonji domain containing 1C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human