Anti-IgG2b Goat Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130792-125EA 2012.84 DKK
ABCAAB130792-125
Anti-IgG2b Goat Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR