Anti-Luteinizing Hormone Rabbit monoclonal antibody [clone: SP132]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130750-100EA 5950 DKK
ABCAAB130750-100
Anti-Luteinizing Hormone Rabbit monoclonal antibody [clone: SP132]
Type: Primary
Antigen: Luteinizing Hormone
Clonality: monoclonal
Clone: SP132
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR