Anti-AFP Rabbit Monoclonal Antibody [clone: SP154]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130748-100EA 4100.24 DKK
ABCAAB130748-100
Anti-AFP Rabbit Monoclonal Antibody [clone: SP154]
Antistoffer
Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: monoclonal
Clone: SP154
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR