Anti-NT5E Mouse Monoclonal Antibody [clone: AD2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130451-100EA 4098.69 DKK
ABCAAB130451-100
Anti-NT5E Mouse Monoclonal Antibody [clone: AD2]
Antistoffer
Type: Primary
Antigen: NT5E (5'-nucleotidase, ecto (CD73))
Clonality: monoclonal
Clone: AD2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR