Anti-ANPEP Mouse Monoclonal Antibody (Biotin) [clone: WM15]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130430-100EA 3951.14 DKK
ABCAAB130430-100
Anti-ANPEP Mouse Monoclonal Antibody (Biotin) [clone: WM15]
Antistoffer
Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: monoclonal
Clone: WM15
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR