Anti-CD55 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-118]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130421-50EA 6440 DKK
ABCAAB130421-50
Anti-CD55 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-118]
Antistoffer
Type: Primary
Antigen: CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))
Clonality: monoclonal
Clone: MEM-118
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR