Anti-SLC38A9 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130398-100EA 3951.14 DKK
ABCAAB130398-100
Anti-SLC38A9 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC38A9 (solute carrier family 38, member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR