Anti-MYLPF Mouse Monoclonal Antibody [clone: MY-38]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130221-100EA 4098.45 DKK
ABCAAB130221-100
Anti-MYLPF Mouse Monoclonal Antibody [clone: MY-38]
Antistoffer
Type: Primary
Antigen: MYLPF (myosin light chain, phosphorylatable, fast skeletal muscle)
Clonality: monoclonal
Clone: MY-38
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR