Anti-TPH1 Mouse Monoclonal Antibody [clone: Try-63]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB130212-100EA 6480 DKK
ABCAAB130212-100
Anti-TPH1 Mouse Monoclonal Antibody [clone: Try-63]
Antistoffer
Type: Primary
Antigen: TPH1 (tryptophan hydroxylase 1)
Clonality: monoclonal
Clone: Try-63
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestil nu