Anti-Slow skeletal Myosin Mouse Monoclonal Antibody [clone: IML-64]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB130179-100EA 6150 DKK
ABCAAB130179-100
Anti-Slow skeletal Myosin Mouse Monoclonal Antibody [clone: IML-64]
Antistoffer
Type: Primary
Antigen: Slow skeletal Myosin
Clonality: monoclonal
Clone: IML-64
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat