Anti-Serotonin transporter Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130130-200EA 6480 DKK
ABCAAB130130-200
Anti-Serotonin transporter Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Serotonin transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR