Anti-VCL Mouse Monoclonal Antibody [clone: VIN-54]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB130007-100EA 6470 DKK
ABCAAB130007-100
Anti-VCL Mouse Monoclonal Antibody [clone: VIN-54]
Antistoffer
Type: Primary
Antigen: VCL (vinculin)
Clonality: monoclonal
Clone: VIN-54
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat