Anti-NEB Mouse Monoclonal Antibody [clone: Neb-20]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB130000-100EA 3949.41 DKK
ABCAAB130000-100
Anti-NEB Mouse Monoclonal Antibody [clone: Neb-20]
Antistoffer
Type: Primary
Antigen: NEB (nebulin)
Clonality: monoclonal
Clone: Neb-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR