Anti-Blood Group Antigen B Mouse Monoclonal Antibody [clone: B460]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129992-500EA 6150 DKK
ABCAAB129992-500
Anti-Blood Group Antigen B Mouse Monoclonal Antibody [clone: B460]
Antistoffer
Type: Primary
Antigen: Blood Group Antigen B
Clonality: monoclonal
Clone: B460
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR