Anti-ANO9 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129744-100EA 6450 DKK
ABCAAB129744-100
Anti-ANO9 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ANO9 (anoctamin 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR