Anti-UGT Mouse Monoclonal Antibody [clone: 7H2AF11]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB129729-100EA 6580 DKK
ABCAAB129729-100
Anti-UGT Mouse Monoclonal Antibody [clone: 7H2AF11]
Antistoffer
Type: Primary
Antigen: UGT
Clonality: monoclonal
Clone: 7H2AF11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu