Anti-FOXN3 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129453-100EA 4100.24 DKK
ABCAAB129453-100
Anti-FOXN3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FOXN3 (forkhead box N3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR