Anti-CBFA2T3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129444-100EA 6480 DKK
ABCAAB129444-100
Anti-CBFA2T3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CBFA2T3 (core-binding factor, runt domain, alpha subunit 2; translocated to, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR