Anti-MELK Mouse Monoclonal Antibody [clone: 2G2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129373-100EA 6630 DKK
ABCAAB129373-100
Anti-MELK Mouse Monoclonal Antibody [clone: 2G2]
Antistoffer
Type: Primary
Antigen: MELK (maternal embryonic leucine zipper kinase)
Clonality: monoclonal
Clone: 2G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR