Anti-CYP39A1 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB129334-50EA 0 DKK
ABCAAB129334-50
Anti-CYP39A1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP39A1 (cytochrome P450, family 39, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human