Anti-RIT2 Mouse Monoclonal Antibody [clone: 27G2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129330-50EA 6150 DKK
ABCAAB129330-50
Anti-RIT2 Mouse Monoclonal Antibody [clone: 27G2]
Antistoffer
Type: Primary
Antigen: RIT2 (Ras-like without CAAX 2)
Clonality: monoclonal
Clone: 27G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR