Anti-SLC16A7 Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129290-50EA 4100.24 DKK
ABCAAB129290-50
Anti-SLC16A7 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC16A7 (solute carrier family 16 (monocarboxylate transporter), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR