Anti-ITM2B Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129282-50EA 3802.04 DKK
ABCAAB129282-50
Anti-ITM2B Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ITM2B (integral membrane protein 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR