Anti-ERLIN2 Rabbit Monoclonal Antibody [clone: EPR8088]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129207-100EA 3354.74 DKK
ABCAAB129207-100
Anti-ERLIN2 Rabbit Monoclonal Antibody [clone: EPR8088]
Antistoffer
Type: Primary
Antigen: ERLIN2 (ER lipid raft associated 2)
Clonality: monoclonal
Clone: EPR8088
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR