Anti-PGAM1 Rabbit Monoclonal Antibody [clone: EPR8042]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129191-100EA 6890 DKK
ABCAAB129191-100
Anti-PGAM1 Rabbit Monoclonal Antibody [clone: EPR8042]
Antistoffer
Type: Primary
Antigen: PGAM1 (phosphoglycerate mutase 1 (brain))
Clonality: monoclonal
Clone: EPR8042
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR