Anti-SHB Rabbit Monoclonal Antibody [clone: EPR7976]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129190-100EA 3913.86 DKK
ABCAAB129190-100
Anti-SHB Rabbit Monoclonal Antibody [clone: EPR7976]
Antistoffer
Type: Primary
Antigen: SHB (Src homology 2 domain containing adaptor protein B)
Clonality: monoclonal
Clone: EPR7976
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR