Anti-NAPSIN A Rabbit Monoclonal Antibody [clone: EPR6257]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129189-100EA 4098.45 DKK
ABCAAB129189-100
Anti-NAPSIN A Rabbit Monoclonal Antibody [clone: EPR6257]
Antistoffer
Type: Primary
Antigen: NAPSIN A
Clonality: monoclonal
Clone: EPR6257
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR