Anti-CTBP1 Rabbit monoclonal antibody unconjugated [clone: EPR6800]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129181-100EA 6690 DKK
ABCAAB129181-100
Anti-CTBP1 Rabbit monoclonal antibody unconjugated [clone: EPR6800]
Antistoffer
Type: Primary
Antigen: CTBP1 (C-terminal binding protein 1)
Clonality: monoclonal
Clone: EPR6800
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR