Anti-HIRA Rabbit Monoclonal Antibody [clone: EPR7416]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129169-100EA 6580 DKK
ABCAAB129169-100
Anti-HIRA Rabbit Monoclonal Antibody [clone: EPR7416]
Antistoffer
Type: Primary
Antigen: HIRA (histone cell cycle regulator)
Clonality: monoclonal
Clone: EPR7416
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR