Anti-AP2B1 Rabbit Monoclonal Antibody [clone: EPR7567]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129168-100EA 6020 DKK
ABCAAB129168-100
Anti-AP2B1 Rabbit Monoclonal Antibody [clone: EPR7567]
Antistoffer
Type: Primary
Antigen: AP2B1 (adaptor-related protein complex 2, beta 1 subunit)
Clonality: monoclonal
Clone: EPR7567
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR