Anti-NCS1 Rabbit Monoclonal Antibody [clone: EPR7699]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB129166-100EA 7130 DKK
ABCAAB129166-100
Anti-NCS1 Rabbit Monoclonal Antibody [clone: EPR7699]
Antistoffer
Type: Primary
Antigen: NCS1 (neuronal calcium sensor 1)
Clonality: monoclonal
Clone: EPR7699
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu