Anti-LSM2 Rabbit Monoclonal Antibody [clone: EPR7216]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129164-100EA 5850 DKK
ABCAAB129164-100
Anti-LSM2 Rabbit Monoclonal Antibody [clone: EPR7216]
Antistoffer
Type: Primary
Antigen: LSM2 (LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae))
Clonality: monoclonal
Clone: EPR7216
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR