Anti-PFDN5 Rabbit Monoclonal Antibody [clone: EPR7755]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB129116-100EA 6020 DKK
ABCAAB129116-100
Anti-PFDN5 Rabbit Monoclonal Antibody [clone: EPR7755]
Antistoffer
Type: Primary
Antigen: PFDN5 (prefoldin subunit 5)
Clonality: monoclonal
Clone: EPR7755
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu