Anti-EPM2A Rabbit Monoclonal Antibody [clone: EP7235]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB129110-100EA 3876.59 DKK
ABCAAB129110-100
Anti-EPM2A Rabbit Monoclonal Antibody [clone: EP7235]
Antistoffer
Type: Primary
Antigen: EPM2A (epilepsy, progressive myoclonus type 2A, Lafora disease (laforin))
Clonality: monoclonal
Clone: EP7235
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR